Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046063.1 | NP_001039528.1 | 5 - 6 | 1425 | 123 | |
GenBank mRNA | BC105338.1 | - | 5 - 6 | 1425 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |