Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046109.2 | NP_001039574.1 | 3 - 4 | 733 | 92 | |
GenBank mRNA | BC110283.1 | - | 3 - 4 | 734 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |