Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046116.2 | NP_001039581.1 | 6 - 7 | 1005 | 54 | |
GenBank mRNA | BC112794.1 | - | 6 - 7 | 1006 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |