Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046230.1 | NP_001039695.1 | 6 - 7 | 525 | 72 | |
GenBank mRNA | BC110249.1 | - | 6 - 7 | 525 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |