Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046245.2 | NP_001039710.1 | 1 - 2 | 304 | 69 | |
GenBank mRNA | BC105451.1 | - | 1 - 2 | 305 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |