Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046255.2 | NP_001039720.1 | 4 - 5 | 594 | 69 | |
GenBank mRNA | BC112769.1 | - | 4 - 5 | 596 | 69 | |
BC151566.1 | - | 4 - 5 | 574 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |