Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046259.2 | NP_001039724.1 | 2 - 3 | 735 | 148 | |
GenBank mRNA | BC110202.1 | - | 2 - 3 | 736 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |