Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046270.2 | NP_001039735.1 | 1 - 2 | 187 | 72 | |
GenBank mRNA | BC112555.1 | - | 1 - 2 | 188 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |