Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046421.2 | NP_001039886.1 | 7 - 8 | 963 | 151 | |
GenBank mRNA | BC112823.1 | - | 7 - 8 | 964 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |