Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075404.1 | NP_001068872.1 | 8 - 9 | 890 | 154 | |
GenBank mRNA | BC111215.1 | - | 8 - 9 | 890 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |