Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076290.2 | NP_001069758.1 | 1 - 2 | 132 | 129 | |
GenBank mRNA | BC109666.1 | - | 1 - 2 | 133 | 129 | |
EU140625.1 | - | 1 - 2 | 192 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |