Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076365.2 | NP_001069833.1 | 6 - 7 | 785 | 148 | |
GenBank mRNA | BC109626.1 | - | 6 - 7 | 787 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |