Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076505.2 | NP_001069973.1 | 3 - 4 | 187 | 162 | |
GenBank mRNA | BC111663.1 | - | 3 - 4 | 188 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |