Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076962.2 | NP_001070430.1 | 4 - 5 | 698 | 115 | |
GenBank mRNA | BC111227.1 | - | 4 - 5 | 699 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |