Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077055.2 | NP_001070523.1 | 1 - 2 | 26 | 64 | |
GenBank mRNA | BC109747.1 | - | 1 - 2 | 27 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |