Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8H8orf74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077126.1 | NP_001070594.1 | 3 - 4 | 667 | 61 | |
GenBank mRNA | BC111241.1 | - | 3 - 4 | 667 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |