Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078038.1 | NP_001071506.1 | 3 - 4 | 446 | 102 | |
GenBank mRNA | BC111261.1 | - | 3 - 4 | 446 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |