Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079618.2 | NP_001073086.1 | 2 - 3 | 479 | 77 | |
GenBank mRNA | BC110250.1 | - | 2 - 3 | 480 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |