Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080357.2 | NP_001073826.1 | 1 - 2 | 453 | 61 | |
GenBank mRNA | BC110224.1 | - | 1 - 2 | 454 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |