Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174635.2 | NP_777060.2 | 6 - 7 | 656 | 147 | |
GenBank mRNA | BC112816.1 | - | 6 - 7 | 656 | 147 | |
M93665.1 | - | 5 - 6 | 556 | 147 | ||
X54962.1 | - | 620 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |