Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015627.2 | NP_001015627.1 | 2 - 3 | 231 | 58 | |
GenBank mRNA | BC118313.1 | - | 2 - 3 | 231 | 58 | |
BT020922.1 | - | 2 - 3 | 189 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |