Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045931.1 | NP_001039396.1 | 5 - 6 | 1511 | 125 | |
GenBank mRNA | BC105450.1 | - | 5 - 6 | 1511 | 125 | |
BT029822.1 | - | 5 - 6 | 1498 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |