Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045961.1 | NP_001039426.1 | 6 - 7 | 1677 | 75 | |
GenBank mRNA | BC114839.1 | - | 6 - 7 | 1677 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |