Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046073.2 | NP_001039538.1 | 3 - 4 | 427 | 83 | |
GenBank mRNA | BC114127.1 | - | 3 - 4 | 428 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |