Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046131.2 | NP_001039596.1 | 5 - 6 | 938 | 85 | |
GenBank mRNA | BC105481.1 | - | 5 - 6 | 939 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |