Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046406.2 | NP_001039871.1 | 1 - 2 | 162 | 67 | |
GenBank mRNA | BC112760.1 | - | 1 - 2 | 163 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |