Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046598.2 | NP_001040063.1 | 2 - 3 | 201 | 107 | |
GenBank mRNA | BC105371.1 | - | 2 - 3 | 202 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |