Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046605.1 | NP_001040070.1 | 4 - 5 | 590 | 68 | |
GenBank mRNA | BC114113.1 | - | 4 - 5 | 590 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |