Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046624.2 | NP_001040089.1 | 1 - 2 | 355 | 64 | |
GenBank mRNA | BC114121.1 | - | 1 - 2 | 356 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |