Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075163.1 | NP_001068631.1 | 1 - 2 | 74 | 78 | |
GenBank mRNA | BC118224.1 | - | 1 - 2 | 74 | 78 | |
BC151415.1 | - | 1 - 2 | 95 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |