Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075163.1 | NP_001068631.1 | 3 - 4 | 288 | 98 | |
GenBank mRNA | BC118224.1 | - | 3 - 4 | 288 | 98 | |
BC151415.1 | - | 3 - 4 | 309 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |