Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075170.1 | NP_001068638.1 | 9 - 10 | 780 | 129 | |
GenBank mRNA | BC114829.1 | - | 9 - 10 | 780 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |