Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075209.1 | NP_001068677.1 | 8 - 9 | 832 | 86 | |
GenBank mRNA | BC116107.1 | - | 8 - 9 | 832 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |