Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075874.1 | NP_001069342.1 | 4 - 5 | 559 | 64 | |
GenBank mRNA | BC118196.1 | - | 4 - 5 | 559 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |