Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076119.1 | NP_001069587.1 | 7 - 8 | 1099 | 82 | |
GenBank mRNA | BC118327.1 | - | 7 - 8 | 1099 | 82 | |
BC120462.1 | - | 4 - 5 | 438 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |