Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076130.2 | NP_001069598.1 | 3 - 4 | 637 | 70 | |
GenBank mRNA | BC114817.1 | - | 3 - 4 | 638 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |