Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076352.1 | NP_001069820.1 | 3 - 4 | 699 | 71 | |
GenBank mRNA | BC116134.1 | - | 3 - 4 | 699 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |