Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076380.2 | NP_001069848.1 | 7 - 8 | 1085 | 75 | |
GenBank mRNA | BC116001.1 | - | 7 - 8 | 1086 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |