Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191119.3 | NP_001178048.1 | 7 - 8 | 821 | 96 | |
GenBank mRNA | BC114156.1 | - | 7 - 8 | 822 | 96 | |
X05717.1 | - | 906 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |