Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001174.2 | NP_001001174.2 | 2 - 3 | 220 | 90 | |
GenBank mRNA | BC122591.1 | - | 2 - 3 | 220 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |