Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075430.2 | NP_001068898.1 | 3 - 4 | 533 | 65 | |
GenBank mRNA | BC120187.1 | - | 3 - 4 | 535 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |