Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075467.2 | NP_001068935.1 | 7 - 8 | 909 | 144 | |
GenBank mRNA | BC120170.1 | - | 7 - 8 | 910 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |