Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075516.1 | NP_001068984.1 | 3 - 4 | 635 | 90 | |
GenBank mRNA | BC120090.1 | - | 3 - 4 | 635 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |