Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075570.1 | NP_001069038.1 | 14 - 15 | 1771 | 135 | |
GenBank mRNA | BT026545.1 | - | 14 - 15 | 1771 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |