Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075725.1 | NP_001069193.1 | 3 - 4 | 381 | 110 | |
GenBank mRNA | BC120050.1 | - | 3 - 4 | 381 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |