Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075844.1 | NP_001069312.1 | 9 - 10 | 1365 | 108 | |
GenBank mRNA | BC123418.1 | - | 9 - 10 | 1365 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |