Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075983.1 | NP_001069451.1 | 1 - 2 | 215 | 89 | |
GenBank mRNA | BC123722.1 | - | 1 - 2 | 215 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |