Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075983.1 | NP_001069451.1 | 5 - 6 | 884 | 78 | |
GenBank mRNA | BC123722.1 | - | 5 - 6 | 884 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |