Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076043.2 | NP_001069511.1 | 2 - 3 | 374 | 110 | |
GenBank mRNA | BC120094.1 | - | 2 - 3 | 375 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |