Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7H5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076110.1 | NP_001069578.1 | 1 - 2 | 215 | 95 | |
GenBank mRNA | BC122833.1 | - | 1 - 2 | 215 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |